Search
Search

AT2G23680| Gene Symbol: AT2G23680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124901.2 | NP_001118373.1 | 1 - 2 | 451 | 80 | |
| NM_127931.3 | NP_179948.1 | 2 - 3 | 359 | 80 | ||
Set Membership: |
|