Search
Search

scpl22| Gene Symbol: scpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084481.1 | NP_001077950.1 | 3 - 4 | 437 | 100 | |
| NM_127962.3 | NP_179978.2 | 5 - 6 | 680 | 100 | ||
| GenBank mRNA | DQ446557.1 | - | 3 - 4 | 437 | 100 | |
| DQ446558.1 | - | 5 - 6 | 626 | 100 | ||
| DQ785742.1 | - | 5 - 6 | 626 | 100 | ||
Set Membership: |
|