Search
Search

SCA3| Gene Symbol: SCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127973.5 | NP_179989.1 | 6 - 7 | 1524 | 96 | |
| NM_201796.2 | NP_973525.2 | 6 - 7 | 1524 | 96 | ||
| GenBank mRNA | AK229652.1 | - | 6 - 7 | 1430 | 96 | |
| Y08463.1 | - | 1471 | 96 | |||
Set Membership: |
|