Search
Search

RAD23B| Gene Symbol: RAD23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198510.1 | NP_001185439.1 | 7 - 8 | 785 | 63 | |
| NM_106614.6 | NP_565216.2 | 6 - 7 | 773 | 63 | ||
| NM_180651.3 | NP_850982.1 | 6 - 7 | 791 | 63 | ||
| NM_202452.2 | NP_974181.1 | 6 - 7 | 703 | 63 | ||
| GenBank mRNA | AY088037.1 | - | 6 - 7 | 703 | 63 | |
Set Membership: |
|