Search
Search

HD2B| Gene Symbol: HD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122171.3 | NP_197657.3 | 8 - 9 | 944 | 83 | |
| NM_180725.3 | NP_851056.1 | 8 - 9 | 1018 | 83 | ||
| GenBank mRNA | AF044914.1 | - | 7 - 8 | 841 | 83 | |
| AF195546.1 | - | 8 - 9 | 901 | 83 | ||
| AY059893.1 | - | 8 - 9 | 918 | 83 | ||
| AY079014.1 | - | 8 - 9 | 916 | 83 | ||
| AY122890.1 | - | 7 - 8 | 844 | 83 | ||
Set Membership: |
|