Search
Search

HCF109| Gene Symbol: HCF109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344128.1 | NP_001332421.1 | 5 - 6 | 929 | 65 | |
| NM_123008.3 | NP_198466.2 | 5 - 6 | 950 | 65 | ||
| NM_180765.2 | NP_851096.1 | 5 - 6 | 978 | 65 | ||
| NM_180766.1 | NP_851097.1 | 5 - 6 | 929 | 65 | ||
| GenBank mRNA | AJ298098.1 | - | 5 - 6 | 877 | 65 | |
| AY056234.1 | - | 5 - 6 | 903 | 65 | ||
| AY084711.1 | - | 5 - 6 | 932 | 65 | ||
| AY117253.1 | - | 5 - 6 | 880 | 65 | ||
Set Membership: |
|