Search
Search

SPDS3| Gene Symbol: SPDS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085279.1 | NP_001078748.1 | 5 - 6 | 496 | 73 | |
| NM_001085280.1 | NP_001078749.1 | 4 - 5 | 401 | 73 | ||
| NM_001203598.1 | NP_001190527.1 | 4 - 5 | 670 | 73 | ||
| NM_001345033.1 | NP_001332707.1 | 4 - 5 | 527 | 73 | ||
| NM_124691.3 | NP_568785.1 | 5 - 6 | 533 | 73 | ||
| NM_180847.2 | NP_851178.1 | 4 - 5 | 593 | 73 | ||
| NM_180848.3 | NP_851179.1 | 4 - 5 | 580 | 73 | ||
| GenBank mRNA | AY040013.1 | - | 4 - 5 | 559 | 73 | |
| AY079367.1 | - | 3 - 4 | 315 | 73 | ||
| BT000742.1 | - | 4 - 5 | 544 | 73 | ||
Set Membership: |
|