Search
Search

FIP2| Gene Symbol: FIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124882.4 | NP_200311.1 | 10 - 11 | 844 | 79 | |
| NM_180862.1 | NP_851193.1 | 10 - 11 | 839 | 79 | ||
| GenBank mRNA | AF174429.1 | - | 10 - 11 | 839 | 79 | |
| BT030093.1 | - | 9 - 10 | 758 | 79 | ||
Set Membership: |
|