Search
Search

ACA8| Gene Symbol: ACA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345242.1 | NP_001331281.1 | 15 - 16 | 1521 | 84 | |
| NM_125093.6 | NP_200521.3 | 15 - 16 | 1570 | 84 | ||
| NM_180869.3 | NP_851200.1 | 15 - 16 | 1412 | 84 | ||
| GenBank mRNA | AJ249352.1 | - | 15 - 16 | 1414 | 84 | |
| AK226961.1 | - | 14 - 15 | 1336 | 84 | ||
| AY069869.1 | - | 15 - 16 | 1414 | 84 | ||
Set Membership: |
|