Search
Search

ACA8| Gene Symbol: ACA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345242.1 | NP_001331281.1 | 22 - 23 | 2105 | 102 | |
| NM_125093.6 | NP_200521.3 | 22 - 23 | 2154 | 102 | ||
| NM_180869.3 | NP_851200.1 | 22 - 23 | 1996 | 102 | ||
| GenBank mRNA | AJ249352.1 | - | 22 - 23 | 1998 | 102 | |
| AK226961.1 | - | 21 - 22 | 1920 | 102 | ||
| AY069869.1 | - | 22 - 23 | 1998 | 102 | ||
Set Membership: |
|