Search
Search

AGL42| Gene Symbol: AGL42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037046.2 | NP_001032123.1 | 5 - 6 | 750 | 153 | |
| NM_001203665.2 | NP_001190594.1 | 4 - 5 | 601 | 153 | ||
| NM_001345529.1 | NP_001330224.1 | 5 - 6 | 765 | 257 | ||
| NM_001345530.1 | NP_001330225.1 | 5 - 6 | 753 | 257 | ||
| NM_001345533.1 | NP_001330219.1 | 4 - 5 | 669 | 153 | ||
| NM_001345535.1 | NP_001330221.1 | 4 - 5 | 509 | 257 | ||
| NM_125610.4 | NP_568952.1 | 5 - 6 | 764 | 153 | ||
| NM_180916.1 | NP_851247.1 | 4 - 5 | 800 | 153 | ||
| GenBank mRNA | AY054220.1 | - | 4 - 5 | 801 | 153 | |
| AY065206.1 | - | 4 - 5 | 801 | 153 | ||
| AY066035.1 | - | 4 - 5 | 425 | 153 | ||
| AY096509.1 | - | 4 - 5 | 425 | 153 | ||
| AY141213.1 | - | 4 - 5 | 425 | 153 | ||
Set Membership: |
|