Search
Search

SAC8| Gene Symbol: SAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115042.3 | NP_190751.2 | 15 - 16 | 1439 | 94 | |
| GenBank mRNA | AY080659.1 | - | 15 - 16 | 1411 | 94 | |
| AY133741.1 | - | 15 - 16 | 1323 | 94 | ||
| AY227251.1 | - | 15 - 16 | 1323 | 94 | ||
| U72504.1 | - | 15 - 16 | 1371 | 94 | ||
Set Membership: |
|