Search
Search

SEX4| Gene Symbol: SEX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035765.2 | NP_001030842.1 | 4 - 5 | 505 | 144 | |
| NM_001339563.1 | NP_001326925.1 | 4 - 5 | 469 | 144 | ||
| NM_001339564.1 | NP_001326924.1 | 1 - 2 | 102 | 144 | ||
| NM_115078.4 | NP_566960.1 | 4 - 5 | 601 | 144 | ||
| GenBank mRNA | AJ302781.1 | - | 4 - 5 | 386 | 144 | |
Set Membership: |
|