Search
Search

SPP2| Gene Symbol: SPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035769.3 | NP_001030846.1 | 3 - 4 | 703 | 80 | |
| NM_001339569.1 | NP_001326826.1 | 2 - 3 | 1022 | 80 | ||
| NM_001339570.1 | NP_001326827.1 | 3 - 4 | 937 | 80 | ||
| NM_001339571.1 | NP_001326824.1 | 3 - 4 | 890 | 80 | ||
| NM_001339572.1 | NP_001326825.1 | 3 - 4 | 632 | 80 | ||
| NM_115094.4 | NP_566964.1 | 4 - 5 | 805 | 80 | ||
| NM_202688.2 | NP_974417.1 | 3 - 4 | 735 | 80 | ||
| GenBank mRNA | AF356816.1 | - | 4 - 5 | 554 | 80 | |
| AK175395.1 | - | 3 - 4 | 703 | 80 | ||
| BT000909.1 | - | 2 - 3 | 463 | 80 | ||
Set Membership: |
|