Search
Search

FK| Gene Symbol: FK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115154.3 | NP_566975.1 | 11 - 12 | 1099 | 75 | |
| NM_202691.1 | NP_974420.1 | 11 - 12 | 1058 | 75 | ||
| GenBank mRNA | AF256535.1 | - | 11 - 12 | 1007 | 75 | |
| AK227189.1 | - | 11 - 12 | 965 | 75 | ||
| AY064964.1 | - | 11 - 12 | 1067 | 75 | ||
| AY133650.1 | - | 11 - 12 | 883 | 75 | ||
Set Membership: |
|