Search
Search

FK| Gene Symbol: FK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115154.3 | NP_566975.1 | 8 - 9 | 815 | 125 | |
| NM_202691.1 | NP_974420.1 | 8 - 9 | 770 | 125 | ||
| GenBank mRNA | AF256535.1 | - | 8 - 9 | 723 | 125 | |
| AK227189.1 | - | 8 - 9 | 681 | 125 | ||
| AY064964.1 | - | 8 - 9 | 783 | 125 | ||
| AY133650.1 | - | 8 - 9 | 599 | 125 | ||
Set Membership: |
|