Search
Search

LOS4| Gene Symbol: LOS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115171.3 | NP_190879.1 | 3 - 4 | 664 | 80 | |
| GenBank mRNA | BT002444.1 | - | 3 - 4 | 615 | 80 | |
| BT008867.1 | - | 2 - 3 | 533 | 80 | ||
Set Membership: |
|