Search
Search

RGD3| Gene Symbol: RGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203156.1 | NP_001190085.1 | 23 - 24 | 5257 | 71 | |
| NM_115288.8 | NP_190996.3 | 22 - 23 | 5288 | 71 | ||
| GenBank mRNA | AB440793.1 | - | 21 - 22 | 5026 | 71 | |
Set Membership: |
|