Search
Search

FBL17| Gene Symbol: FBL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115322.4 | NP_567005.2 | 3 - 4 | 1200 | 133 | |
| GenBank mRNA | AF462827.1 | - | 3 - 4 | 894 | 133 | |
| AY142006.1 | - | 3 - 4 | 868 | 133 | ||
Set Membership: |
|