Search
Search

MRH2| Gene Symbol: MRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339702.1 | NP_001326426.1 | 14 - 15 | 2390 | 116 | |
| NM_001339703.1 | NP_001326427.1 | 14 - 15 | 2349 | 116 | ||
| NM_115344.4 | NP_191047.3 | 14 - 15 | 2264 | 116 | ||
| GenBank mRNA | AB290928.1 | - | 14 - 15 | 2279 | 116 | |
| EU257694.1 | - | 14 - 15 | 2264 | 116 | ||
Set Membership: |
|