Search
Search

MRH2| Gene Symbol: MRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339702.1 | NP_001326426.1 | 3 - 4 | 651 | 86 | |
| NM_001339703.1 | NP_001326427.1 | 3 - 4 | 613 | 86 | ||
| NM_115344.4 | NP_191047.3 | 3 - 4 | 525 | 86 | ||
| GenBank mRNA | AB290928.1 | - | 3 - 4 | 540 | 86 | |
| EU257694.1 | - | 3 - 4 | 525 | 86 | ||
Set Membership: |
|