Search
Search

TT5| Gene Symbol: TT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115370.4 | NP_191072.1 | 1 - 2 | 318 | 95 | |
| GenBank mRNA | AJ418046.1 | - | 1 - 2 | 187 | 95 | |
| BT004265.1 | - | 1 - 2 | 187 | 95 | ||
| BT005528.1 | - | 1 - 2 | 136 | 95 | ||
Set Membership: |
|