Search
Search

AT3G55240| Gene Symbol: AT3G55240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115382.5 | NP_191084.1 | 2 - 3 | 770 | 80 | |
| GenBank mRNA | AY080704.1 | - | 2 - 3 | 196 | 80 | |
| BT000964.1 | - | 2 - 3 | 143 | 80 | ||
Set Membership: |
|