Search
Search

AT3G55890| Gene Symbol: AT3G55890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339757.1 | NP_001325435.1 | 3 - 4 | 550 | 150 | |
| NM_001339758.1 | NP_001325434.1 | 3 - 4 | 422 | 150 | ||
| NM_001339759.1 | NP_001325433.1 | 3 - 4 | 422 | 118 | ||
| NM_115447.2 | NP_191148.1 | 3 - 4 | 550 | 118 | ||
| GenBank mRNA | AK229138.1 | - | 3 - 4 | 341 | 118 | |
| AY050951.1 | - | 3 - 4 | 343 | 118 | ||
| AY091443.1 | - | 3 - 4 | 252 | 118 | ||
Set Membership: |
|