Search
Search

PCME| Gene Symbol: PCME | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121591.5 | NP_197090.2 | 1 - 2 | 774 | 92 | |
| NM_203057.1 | NP_974786.1 | 1 - 2 | 303 | 92 | ||
| GenBank mRNA | AY045829.1 | - | 1 - 2 | 395 | 92 | |
| AY091367.1 | - | 1 - 2 | 276 | 92 | ||
Set Membership: |
|