Search
Search

AT5G16300| Gene Symbol: AT5G16300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036813.2 | NP_001031890.1 | 3 - 4 | 2703 | 91 | |
| NM_001203391.1 | NP_001190320.1 | 3 - 4 | 2848 | 91 | ||
| NM_121635.4 | NP_197134.1 | 3 - 4 | 2927 | 91 | ||
| NM_203059.2 | NP_974788.1 | 3 - 4 | 2703 | 91 | ||
Set Membership: |
|