Search
Search

DEG8| Gene Symbol: DEG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123346.5 | NP_568575.1 | 1 - 2 | 249 | 92 | |
| NM_203134.2 | NP_974863.1 | 1 - 2 | 216 | 92 | ||
| GenBank mRNA | AY056381.1 | - | 1 - 2 | 193 | 92 | |
| AY090319.1 | - | 1 - 2 | 130 | 92 | ||
Set Membership: |
|