Search
Search

ARP9| Gene Symbol: ARP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344487.1 | NP_001331886.1 | 3 - 4 | 681 | 140 | |
| NM_001344488.1 | NP_001331887.1 | 3 - 4 | 523 | 140 | ||
| NM_123716.3 | NP_199163.1 | 3 - 4 | 635 | 140 | ||
| NM_203147.2 | NP_974876.1 | 3 - 4 | 681 | 140 | ||
| GenBank mRNA | AY057568.1 | - | 3 - 4 | 450 | 140 | |
Set Membership: |
|