Search
Search

PMR5| Gene Symbol: PMR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125247.4 | NP_200668.1 | 1 - 2 | 510 | 80 | |
| NM_203232.2 | NP_974961.1 | 1 - 2 | 484 | 80 | ||
| GenBank mRNA | AK117382.1 | - | 1 - 2 | 394 | 80 | |
| BT005096.1 | - | 1 - 2 | 358 | 80 | ||
Set Membership: |
|