Search
Search

SEN2| Gene Symbol: SEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125416.4 | NP_200831.1 | 2 | 978 | 144 | |
| NM_203237.1 | NP_974966.1 | 1 | 1332 | 144 | ||
| GenBank mRNA | BT004112.1 | - | 2 | 978 | 144 | |
| BT004920.1 | - | 1 | 674 | 144 | ||
Set Membership: |
|