Search
Search

AT1G34320| Gene Symbol: AT1G34320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333096.1 | NP_001322690.1 | 2 - 3 | 1260 | 76 | |
| NM_103154.4 | NP_174692.1 | 3 - 4 | 1184 | 76 | ||
| GenBank mRNA | AK176401.1 | - | 2 - 3 | 715 | 76 | |
| AK176473.1 | - | 3 - 4 | 1032 | 76 | ||
| BT010615.1 | - | 2 - 3 | 644 | 76 | ||
Set Membership: |
|