Search
Search

Lhca5| Gene Symbol: Lhca5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103597.5 | NP_175137.1 | 4 - 5 | 680 | 69 | |
| NM_179447.2 | NP_849778.1 | 4 - 5 | 724 | 69 | ||
| GenBank mRNA | AK221694.1 | - | 4 - 5 | 650 | 69 | |
| AY062097.1 | - | 4 - 5 | 626 | 69 | ||
| AY090263.1 | - | 4 - 5 | 621 | 69 | ||
Set Membership: |
|