Search
Search

KU80| Gene Symbol: KU80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333292.1 | NP_001322094.1 | 4 - 5 | 364 | 113 | |
| NM_001333293.1 | NP_001322093.1 | 3 - 4 | 292 | 113 | ||
| NM_103701.2 | NP_564520.1 | 4 - 5 | 412 | 113 | ||
| GenBank mRNA | AF283758.1 | - | 4 - 5 | 342 | 113 | |
Set Membership: |
|