Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CHR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333322.1 | NP_001321270.1 | 11 - 12 | 1083 | 97 | |
NM_001333323.1 | NP_001321271.1 | 11 - 12 | 960 | 97 | ||
NM_001333324.1 | NP_001321269.1 | 11 - 12 | 1062 | 97 | ||
NM_103728.5 | NP_175265.3 | 11 - 12 | 1068 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |