Search
Search

XIJ| Gene Symbol: XIJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339909.1 | NP_001319790.1 | 16 - 17 | 2168 | 103 | |
| NM_001339910.1 | NP_001327763.1 | 16 - 17 | 2329 | 103 | ||
| NM_001339911.1 | NP_001327764.1 | 16 - 17 | 2329 | 103 | ||
| NM_001339912.1 | NP_001327762.1 | 16 - 17 | 2128 | 103 | ||
| NM_115678.2 | NP_191375.1 | 15 - 16 | 1817 | 103 | ||
Set Membership: |
|