Search
Search

XIJ| Gene Symbol: XIJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339909.1 | NP_001319790.1 | 10 - 11 | 1580 | 93 | |
| NM_001339910.1 | NP_001327763.1 | 10 - 11 | 1741 | 93 | ||
| NM_001339911.1 | NP_001327764.1 | 10 - 11 | 1741 | 93 | ||
| NM_001339912.1 | NP_001327762.1 | 10 - 11 | 1540 | 93 | ||
| NM_115678.2 | NP_191375.1 | 9 - 10 | 1229 | 93 | ||
Set Membership: |
|