Search
Search

RBL15| Gene Symbol: RBL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203197.1 | NP_001190126.1 | 3 - 4 | 315 | 90 | |
| NM_115708.4 | NP_567064.1 | 2 - 3 | 448 | 90 | ||
| GenBank mRNA | AY136386.1 | - | 2 - 3 | 282 | 90 | |
| BT000186.1 | - | 2 - 3 | 187 | 90 | ||
Set Membership: |
|