Search
Search

PAO3| Gene Symbol: PAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115767.3 | NP_191464.1 | 8 - 9 | 1502 | 98 | |
| GenBank mRNA | AY065025.1 | - | 8 - 9 | 1484 | 98 | |
| AY143905.1 | - | 8 - 9 | 1166 | 98 | ||
Set Membership: |
|