Search
Search

GCN2| Gene Symbol: GCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203206.1 | NP_001190135.1 | 18 - 19 | 3046 | 88 | |
| NM_001339969.1 | NP_001326041.1 | 18 - 19 | 3913 | 88 | ||
| NM_001339970.1 | NP_001326042.1 | 17 - 18 | 3055 | 88 | ||
| NM_001339971.1 | NP_001326040.1 | 17 - 18 | 2826 | 88 | ||
| NM_115803.3 | NP_191500.2 | 17 - 18 | 2945 | 88 | ||
| GenBank mRNA | AJ459823.1 | - | 17 - 18 | 2593 | 88 | |
Set Membership: |
|