Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: GCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203206.1 | NP_001190135.1 | 3 - 4 | 659 | 62 | |
NM_001339969.1 | NP_001326041.1 | 3 - 4 | 1526 | 62 | ||
NM_001339970.1 | NP_001326042.1 | 2 - 3 | 665 | 62 | ||
NM_001339971.1 | NP_001326040.1 | 2 - 3 | 439 | 62 | ||
NM_115803.3 | NP_191500.2 | 2 - 3 | 558 | 62 | ||
GenBank mRNA | AJ459823.1 | - | 2 - 3 | 206 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |