Search
Search

VAP27-1| Gene Symbol: VAP27-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035826.1 | NP_001030903.1 | 6 - 7 | 1137 | 82 | |
| NM_001035827.1 | NP_001030904.1 | 6 - 7 | 1157 | 82 | ||
| NM_115924.4 | NP_567101.1 | 7 - 8 | 1085 | 82 | ||
| GenBank mRNA | AY065144.1 | - | 7 - 8 | 796 | 82 | |
| AY364005.1 | - | 7 - 8 | 674 | 82 | ||
| BT006297.1 | - | 7 - 8 | 658 | 82 | ||
Set Membership: |
|