Search
Search

PBF1| Gene Symbol: PBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203217.1 | NP_001190146.1 | 6 - 7 | 755 | 101 | |
| NM_115946.3 | NP_191641.1 | 6 - 7 | 723 | 101 | ||
| GenBank mRNA | AF043537.1 | - | 6 - 7 | 678 | 101 | |
| AY062790.1 | - | 6 - 7 | 692 | 101 | ||
| AY081571.1 | - | 6 - 7 | 593 | 101 | ||
| AY086619.1 | - | 6 - 7 | 713 | 101 | ||
| X79806.1 | - | 652 | 101 | |||
Set Membership: |
|