Search
Search

PEX11E| Gene Symbol: PEX11E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084853.2 | NP_001078322.1 | 8 - 9 | 842 | 85 | |
| NM_001203220.1 | NP_001190149.1 | 7 - 8 | 1009 | 85 | ||
| NM_115971.3 | NP_191666.2 | 7 - 8 | 931 | 85 | ||
| GenBank mRNA | BT002962.1 | - | 7 - 8 | 900 | 85 | |
| BT004439.1 | - | 6 - 7 | 657 | 85 | ||
Set Membership: |
|