Search
Search

HDG1| Gene Symbol: HDG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115979.5 | NP_191674.1 | 5 - 6 | 1726 | 56 | |
| GenBank mRNA | AY050866.1 | - | 5 - 6 | 1447 | 56 | |
| AY096757.1 | - | 5 - 6 | 1288 | 56 | ||
Set Membership: |
|