Search
Search

U2B''| Gene Symbol: U2B'' | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336249.1 | NP_001324159.1 | 3 - 4 | 574 | 101 | |
| NM_128579.4 | NP_180585.1 | 3 - 4 | 574 | 101 | ||
| GenBank mRNA | AK227609.1 | - | 3 - 4 | 418 | 101 | |
| BT003068.1 | - | 3 - 4 | 343 | 101 | ||
Set Membership: |
|