Search
Search

5-FCL| Gene Symbol: 5-FCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036796.2 | NP_001031873.1 | 3 - 4 | 550 | 75 | |
| NM_121308.2 | NP_568284.1 | 3 - 4 | 621 | 75 | ||
| GenBank mRNA | AF516365.1 | - | 3 - 4 | 540 | 75 | |
| AY084396.1 | - | 3 - 4 | 621 | 75 | ||
| BT024502.1 | - | 3 - 4 | 540 | 75 | ||
Set Membership: |
|