Search
Search

FY| Gene Symbol: FY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343274.1 | NP_001318555.1 | 7 - 8 | 951 | 113 | |
| NM_001343275.1 | NP_001331012.1 | 7 - 8 | 882 | 113 | ||
| NM_001343276.1 | NP_001331011.1 | 7 - 8 | 882 | 113 | ||
| NM_121351.5 | NP_196852.3 | 6 - 7 | 918 | 113 | ||
| GenBank mRNA | BT012225.1 | - | 6 - 7 | 645 | 113 | |
Set Membership: |
|