Search
Search

SMT1| Gene Symbol: SMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085110.1 | NP_001078579.1 | 12 - 13 | 1043 | 73 | |
| NM_121374.4 | NP_196875.1 | 13 - 14 | 1327 | 73 | ||
| GenBank mRNA | AF090372.1 | - | 13 - 14 | 994 | 73 | |
| AF195648.1 | - | 13 - 14 | 1005 | 73 | ||
| AY120716.1 | - | 13 - 14 | 1019 | 73 | ||
| BT000058.1 | - | 12 - 13 | 952 | 73 | ||
Set Membership: |
|