Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203382.2 | NP_001190311.1 | 5 - 6 | 858 | 133 | |
NM_121506.4 | NP_197006.2 | 5 - 6 | 850 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |