Search
Search

HB-3| Gene Symbol: HB-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121519.3 | NP_568309.2 | 2 - 3 | 981 | 78 | |
| GenBank mRNA | AK229364.1 | - | 2 - 3 | 897 | 78 | |
| BT044059.1 | - | 2 - 3 | 614 | 78 | ||
Set Membership: |
|